Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11bR)-N-(2,6-Bis(4-methoxyphenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: AMBEED, INC
Warning

A1258444-100MG A1258444-1G A1258444-250MG A1258444-50MG
76680-728EA 68 USD InStock
76680-728 76680-730 76680-732 76680-734
(11bR)-N-(2,6-Bis(4-methoxyphenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bR)-N-(2,6-Bis(4-methoxyphenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide
Precaución: for Research Use Only


Formula: C₃₅H₃₃F₃NO₇PS
MDL Number: MFCD34563599
CAS Number: 1071853-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty