Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibromobis(pyridine)nickel 97%

Supplier: AMBEED, INC
Danger

A1258467-1G A1258467-250MG A1258467-5G
76680-768EA 49 USD InStock
76680-768 76680-770 76680-772
Dibromobis(pyridine)nickel 97%
Dibromobis(pyridine)nickel
Precaución: for Research Use Only


Formula: C₁₀H₁₀Br₂N₂Ni
CAS Number: 14024-85-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty