Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4,4′,4′′,4′′′-Tetraaminophthalocyaninato)cobalt 95%

Supplier: AMBEED, INC
Warning

A1262271-100MG A1262271-250MG A1262271-25MG A1262271-50MG
76680-858EA 101 USD InStock
76680-858 76680-860 76680-862 76680-864
(4,4′,4′′,4′′′-Tetraaminophthalocyaninato)cobalt 95%
(4,4′,4′′,4′′′-Tetraaminophthalocyaninato)cobalt
Precaución: for Research Use Only


Formula: C₃₂H₂₀CoN₁₂
CAS Number: 27680-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty