Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-(Propane-1,3-diyl)diacrylamide 97% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1262898-100MG A1262898-1G A1262898-250MG
76680-874EA 67 USD InStock
76680-874 76680-876 76680-878
N,N'-(Propane-1,3-diyl)diacrylamide 97% stabilized
N,N'-(Propane-1,3-diyl)diacrylamide
Precaución: for Research Use Only


Formula: C₉H₁₄N₂O₂
MDL Number: MFCD00081200
CAS Number: 4887-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty