Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dibromo-6-chloro-1H-benzo[d][1,2,3]triazole 95%

Supplier: AMBEED, INC
Warning

A1263338-100MG A1263338-1G A1263338-250MG
76680-908EA 182 USD InStock
76680-908 76680-910 76680-912
4,7-Dibromo-6-chloro-1H-benzo[d][1,2,3]triazole 95%
4,7-Dibromo-6-chloro-1H-benzo[d][1,2,3]triazole
Precaución: for Research Use Only


Formula: C₆H₂Br₂ClN₃
MDL Number: MFCD31539968
CAS Number: 1086837-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty