Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-((S)-(2-(Dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 98%

Supplier: AMBEED, INC
Warning

A1264532-100MG A1264532-250MG A1264532-50MG
76681-026EA 131 USD InStock
76681-026 76681-028 76681-030
(R)-N-((S)-(2-(Dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 98%
(R)-N-((S)-(2-(Dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide
Precaución: for Research Use Only


Formula: C₂₉H₄₂NOPS
MDL Number: MFCD32697194
CAS Number: 1595319-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty