Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(piperazin-1-ylmethyl)piperidine-1-carboxylate 98%

Supplier: AMBEED, INC
Danger

A1264615-10G A1264615-1G A1264615-250MG A1264615-5G
76681-038EA 90 USD InStock
76681-038 76681-040 76681-042 76681-044
tert-Butyl 4-(piperazin-1-ylmethyl)piperidine-1-carboxylate 98%
tert-Butyl 4-(piperazin-1-ylmethyl)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₅H₂₉N₃O₂
MDL Number: MFCD09753909
CAS Number: 381722-48-1
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty