Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Quinolin-2-yl)propan-2-one 97%

Supplier: AMBEED, INC
Warning

A1271187-100MG A1271187-1G A1271187-250MG
76681-132EA 21 USD InStock
76681-132 76681-134 76681-136
1-(Quinolin-2-yl)propan-2-one 97%
1-(Quinolin-2-yl)propan-2-one
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO
MDL Number: MFCD00552685
CAS Number: 1531-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty