Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibromo(1,10-phenanthroline-κN1,κN10)nickel 98%

Supplier: AMBEED, INC
Warning

A1272891-100G A1272891-1G A1272891-25G A1272891-5G
76681-246EA 960 USD InStock
76681-246 76681-248 76681-250 76681-252
Dibromo(1,10-phenanthroline-κN1,κN10)nickel 98%
Dibromo(1,10-phenanthroline-κN1,κN10)nickel
Precaución: for Research Use Only


Formula: C₁₂H₈Br₂N₂Ni
CAS Number: 48165-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty