Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(7-Acetamido-1H-indole-2-carboxamido)-4-methylpentanoic acid 97%

Supplier: AMBEED, INC
Warning

A1273143-100MG A1273143-1G A1273143-250MG A1273143-25MG A1273143-50MG
76681-272EA 80 USD InStock
76681-272 76681-274 76681-276 76681-278 76681-280
(S)-2-(7-Acetamido-1H-indole-2-carboxamido)-4-methylpentanoic acid 97%
(S)-2-(7-Acetamido-1H-indole-2-carboxamido)-4-methylpentanoic acid
Precaución: for Research Use Only


Formula: C₁₇H₂₁N₃O₄
MDL Number: MFCD32856692
CAS Number: 2682097-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty