Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanophenyl trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Danger

A1274366-100MG A1274366-1G A1274366-250MG A1274366-25G A1274366-5G
76681-322EA 9 USD InStock
76681-322 76681-324 76681-326 76681-328 76681-330
4-Cyanophenyl trifluoromethanesulfonate 98%
4-Cyanophenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₈H₄F₃NO₃S
MDL Number: MFCD00973310
CAS Number: 66107-32-2
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty