Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(5-((Pyridin-3-ylmethyl)amino)pyrazolo[1,5-a]pyrimidin-3-yl)benzamide 98+%

Supplier: AMBEED, INC
Warning

A1275491-10MG A1275491-1MG A1275491-50MG A1275491-5MG
76681-404EA 59 USD InStock
76681-404 76681-406 76681-408 76681-410
4-(5-((Pyridin-3-ylmethyl)amino)pyrazolo[1,5-a]pyrimidin-3-yl)benzamide 98+%
4-(5-((Pyridin-3-ylmethyl)amino)pyrazolo[1,5-a]pyrimidin-3-yl)benzamide
Precaución: for Research Use Only


Formula: C₁₉H₁₆N₆O
MDL Number: MFCD32693930
CAS Number: 2414909-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty