Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-5-methoxypicolinate 97%

Supplier: AMBEED, INC
Warning

A1275621-100MG A1275621-250MG A1275621-1G
76681-416EA 81 USD InStock
76681-416 76681-418 77621-442
Methyl 4-bromo-5-methoxypicolinate 97%
Methyl 4-bromo-5-methoxypicolinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₃
MDL Number: MFCD18257510
CAS Number: 1256803-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty