Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

14-Bromotetradecanoic acid 98%

Supplier: AMBEED, INC
Warning

A1275891-250MG A1275891-5G A1275891-1G A1275891-100MG
76681-432EA 45 USD InStock
76681-432 76681-434 77592-486 77621-444
14-Bromotetradecanoic acid 98%
14-Bromotetradecanoic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₇BrO₂
MW: 307.2670 g/mol
MDL Number: MFCD31807218
CAS Number: 676-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty