Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dihydroxy-3-nitroanthraquinone 98%

Supplier: AMBEED, INC
Warning

A1279704-100MG A1279704-1G A1279704-250MG
76681-476EA 61 USD InStock
76681-476 76681-478 76681-480
1,2-Dihydroxy-3-nitroanthraquinone 98%
1,2-Dihydroxy-3-nitroanthraquinone
Precaución: for Research Use Only


Formula: C₁₄H₇NO₆
MDL Number: MFCD00019145
CAS Number: 568-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty