Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 98%

Supplier: AMBEED, INC
Warning

A1281250-100MG A1281250-1G A1281250-250MG
76681-528EA 140 USD InStock
76681-528 76681-530 76681-532
4,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 98%
4,6-Bis((4S,5S)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan
Precaución: for Research Use Only


Formula: C₄₂H₃₀N₂O₃
CAS Number: 2067322-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty