Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dimethylcyclohexanone 97%, (Chunks or pellets)

Supplier: AMBEED, INC
Danger

A1320878-25G A1320878-5G
76681-638EA 31 USD InStock
76681-638 76681-640
4,4-Dimethylcyclohexanone 97%, (Chunks or pellets)
4,4-Dimethylcyclohexanone
Precaución: for Research Use Only


Formula: C₈H₁₄O
MDL Number: MFCD00234965
CAS Number: 4255-62-3
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty