Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Amino-2-methylpyrimidin-5-yl)methanol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A1320982-100MG A1320982-250MG A1320982-5G
76681-664EA 18 USD InStock
76681-664 76681-666 76681-668
(4-Amino-2-methylpyrimidin-5-yl)methanol hydrochloride 98%
(4-Amino-2-methylpyrimidin-5-yl)methanol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₀ClN₃O
MDL Number: MFCD27966926
CAS Number: 13100-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty