Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,5R)-1-Hydroxy-2,5-diphenylphospholane 1-oxide 98%

Supplier: AMBEED, INC
Warning

A1326423-100MG A1326423-1G A1326423-5G
76681-798EA 78 USD InStock
76681-798 76681-800 76681-802 76681-804
(2R,5R)-1-Hydroxy-2,5-diphenylphospholane 1-oxide 98%
(2R,5R)-1-Hydroxy-2,5-diphenylphospholane 1-oxide
Precaución: for Research Use Only


Formula: C₁₆H₁₇O₂P
CAS Number: 226393-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty