Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-amino-6-chloro-5-fluoronicotinate 97%

Supplier: AMBEED, INC
Warning

A1327729-10G A1327729-5G A1327729-100MG
76681-870EA 176 USD InStock
76681-870 76681-872 77621-540
Ethyl 4-amino-6-chloro-5-fluoronicotinate 97%
Ethyl 4-amino-6-chloro-5-fluoronicotinate
Precaución: for Research Use Only


Formula: C₈H₈ClFN₂O₂
MDL Number: MFCD34471315
CAS Number: 2454397-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty