Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(1,10-Phenanthroline-2,9-diyl)dianiline 97%

Supplier: AMBEED, INC
Warning

A1327909-100MG A1327909-250MG A1327909-1G
76681-876EA 83 USD InStock
76681-876 76681-878 77621-542
4,4'-(1,10-Phenanthroline-2,9-diyl)dianiline 97%
4,4'-(1,10-Phenanthroline-2,9-diyl)dianiline
Precaución: for Research Use Only


Formula: C₂₄H₁₈N₄
CAS Number: 861659-70-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty