Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl Hydrogen Phosphate 98%

Supplier: AMBEED, INC
Warning

A1340787-100MG A1340787-1G A1340787-250MG
76682-044EA 139 USD InStock
76682-044 76682-046 76682-048
(S)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl Hydrogen Phosphate 98%
(S)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl Hydrogen Phosphate
Precaución: for Research Use Only


Formula: C₄₈H₃₇O₄P
CAS Number: 2099033-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty