Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzo[b,e][1,4]dioxine-2,3,7,8-tetraamine tetrahydrochloride 97%

Supplier: AMBEED, INC
Warning

A1351770-100MG A1351770-1G A1351770-250MG A1351770-5G
76682-106EA 85 USD InStock
76682-106 76682-108 76682-110 76682-112
Dibenzo[b,e][1,4]dioxine-2,3,7,8-tetraamine tetrahydrochloride 97%
Dibenzo[b,e][1,4]dioxine-2,3,7,8-tetraamine tetrahydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₆Cl₄N₄O₂
MDL Number: MFCD32670692
CAS Number: 16435-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty