Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Bis((R)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%

Supplier: AMBEED, INC
Warning

A1351970-100MG A1351970-1G A1351970-250MG
76682-182EA 63 USD InStock
76682-182 76682-184 77621-654
2,6-Bis((R)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
2,6-Bis((R)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only


Formula: C₂₃H₃₁N₃O₂
CAS Number: 2471850-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty