Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-6,6'-Bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphino)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine 98+%

Supplier: AMBEED, INC
Warning

A1386680-100MG A1386680-1G A1386680-250MG A1386680-50MG
76682-464EA 160 USD InStock
76682-464 76682-466 76682-468 77622-006
(S)-6,6'-Bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphino)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine 98+%
(S)-6,6'-Bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphino)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine
Precaución: for Research Use Only


Formula: C₆₄H₉₆O₄P₂Si₈
CAS Number: 2757287-32-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty