Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A653585-100MG A653585-1G A653585-250MG
76682-490EA 161 USD InStock
76682-490 76682-492 77622-420
2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(4-Ethoxy-3-ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₆H₂₅BO₃
MDL Number: MFCD30344676
CAS Number: 2121514-94-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty