Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Roflumilast N-oxide 98%

Supplier: AMBEED, INC
Warning

A981483-10MG A981483-1MG A981483-25MG A981483-2MG A981483-5MG
76682-528EA 119 USD InStock
76682-528 76682-530 76682-532 76682-534 76682-536
Roflumilast N-oxide 98%
Roflumilast N-oxide
Precaución: for Research Use Only


Formula: C₁₇H₁₄Cl₂F₂N₂O₄
MDL Number: MFCD04112984
CAS Number: 292135-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty