Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,4-Difluorophenyl)-5-methylpyridine 97%

Supplier: AMBEED, INC
Warning

A983643-100MG A983643-250MG A983643-1G
76682-540EA 8 USD InStock
76682-540 76682-542 77569-706
2-(2,4-Difluorophenyl)-5-methylpyridine 97%
2-(2,4-Difluorophenyl)-5-methylpyridine
Precaución: For research use only.


Formula: C₁₂H₉F₂N
MDL Number: MFCD17170319
CAS Number: 583052-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty