Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-5-tert-butyl-2-methoxybenzene 95%

Supplier: AMBEED, INC
Warning

A988775-1G A988775-25G A988775-5G
76682-578EA 6 USD InStock
76682-578 76682-580 76682-582
1,3-Dibromo-5-tert-butyl-2-methoxybenzene 95%
1,3-Dibromo-5-tert-butyl-2-methoxybenzene
Precaución: for Research Use Only


Formula: C₁₁H₁₄Br₂O
MW: 322.0363 g/mol
MDL Number: MFCD00233136
CAS Number: 132268-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty