Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(S-Methylsulfonimidoyl)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A994995-100MG A994995-250MG
76682-654EA 103 USD InStock
76682-654 77623-434
4-(S-Methylsulfonimidoyl)benzonitrile 95%
4-(S-Methylsulfonimidoyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD31706254
CAS Number: 1621962-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty