Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate 95%

Supplier: AMBEED, INC
Warning

A995926-1G A995926-250MG A995926-5G
76682-666EA 117 USD InStock
76682-666 76682-668 76682-670
Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate 95%
Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate
Precaución: for Research Use Only


Formula: C₁₈H₂₂BNO₄
MDL Number: MFCD20527090
CAS Number: 2828447-15-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty