Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Adipic acid monobenzyl ester 97%

Supplier: AMBEED, INC
Warning

A100565-1G A100565-5G
76687-002EA 98 USD InStock
76687-002 77623-814
Adipic acid monobenzyl ester 97%
Adipic acid monobenzyl ester
Precaución: For research use only.


Formula: C₁₃H₁₆O₄
MDL Number: MFCD00441728
CAS Number: 40542-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty