Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentanecarbonyl chloride 98%

Supplier: AMBEED, INC
Danger

A101710-1G A101710-25G A101710-5G
76687-048EA 18 USD InStock
76687-048 76687-050 76687-052
Cyclopentanecarbonyl chloride 98%
Cyclopentanecarbonyl chloride
Precaución: for Research Use Only


Formula: C₆H₉ClO
MW: 132.59 g/mol
MDL Number: MFCD00001370
CAS Number: 4524-93-0
UN: 2920
ADR: 8,3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty