Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aprepitant 98%

Supplier: AMBEED, INC
Warning

A103179-10MG A103179-1G A103179-50MG A103179-5G A103179-5MG
76687-110EA 6 USD InStock
76687-110 76687-112 76687-114 76687-116 76687-118
Aprepitant 98%
Aprepitant
Precaución: for Research Use Only


Formula: C₂₃H₂₁F₇N₄O₃
MDL Number: MFCD08277635
CAS Number: 170729-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty