Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Nitroquinoline 98%

Supplier: AMBEED, INC
Warning

A104022-100G A104022-10G A104022-25G A104022-500G A104022-5G
76687-154EA 103 USD InStock
76687-154 76687-156 76687-158 76687-160 76687-162
8-Nitroquinoline 98%
8-Nitroquinoline
Precaución: for Research Use Only


Formula: C₉H₆N₂O₂
MDL Number: MFCD00006806
CAS Number: 607-35-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty