Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloride ionophore I 97%

Supplier: AMBEED, INC
Warning

A105016-100MG A105016-250MG
76687-210EA 31 USD InStock
76687-210 76687-212
Chloride ionophore I 97%
Chloride ionophore I
Precaución: for Research Use Only


Formula: C₄₄H₂₈ClMnN₄
MW: 703.11 g/mol
MDL Number: MFCD00012153
CAS Number: 32195-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty