Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyldimethylphenylammonium chloride 98%

Supplier: AMBEED, INC
Danger

A105271-25G A105271-5G
76687-226EA 37 USD InStock
76687-226 76687-228
Benzyldimethylphenylammonium chloride 98%
Benzyldimethylphenylammonium chloride
Precaución: for Research Use Only


Formula: C₁₅H₁₈ClN
MW: 247.77 g/mol
MDL Number: MFCD00031566
CAS Number: 3204-68-0
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty