Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Methoxyphenyl)glycine 95%

Supplier: AMBEED, INC
Warning

A108402-100MG A108402-10G A108402-1G A108402-250MG
76687-338EA 15 USD InStock
76687-338 76687-340 76687-342 76687-344
2-(2-Methoxyphenyl)glycine 95%
2-(2-Methoxyphenyl)glycine
Precaución: for Research Use Only


Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
MDL Number: MFCD02662408
CAS Number: 271583-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty