Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-methylbenzothiazole 98%

Supplier: AMBEED, INC
Warning

A108777-100G A108777-25G A108777-500G A108777-5G
76687-378EA 25 USD InStock
76687-378 76687-380 76687-382 76687-384
2-Amino-4-methylbenzothiazole 98%
2-Amino-4-methylbenzothiazole
Precaución: for Research Use Only


Formula: C₈H₈N₂S
MDL Number: MFCD00005793
CAS Number: 1477-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty