Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutylmethanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A109029-100G A109029-1G A109029-25G A109029-5G
76687-398EA 399 USD InStock
76687-398 76687-400 76687-402 76687-404
Cyclobutylmethanamine hydrochloride 97%
Cyclobutylmethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClN
MW: 121.6100 g/mol
MDL Number: MFCD00034952
CAS Number: 5454-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty