Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroadenosine hemihydrate 97%

Supplier: AMBEED, INC
Warning

A110823-10G A110823-1G A110823-250MG A110823-5G
76687-556EA 64 USD InStock
76687-556 76687-558 76687-560 76687-562
2-Chloroadenosine hemihydrate 97%
2-Chloroadenosine hemihydrate
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClN₅O₄·0.5H₂O
MDL Number: MFCD00149351
CAS Number: 81012-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty