Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Nitrophenyl)piperazine 98%

Supplier: AMBEED, INC
Warning

A111058-100G A111058-25G A111058-5G
76687-584EA 63 USD InStock
76687-584 76687-586 76687-588
1-(4-Nitrophenyl)piperazine 98%
1-(4-Nitrophenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD00005961
CAS Number: 6269-89-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty