Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid 95%

Supplier: AMBEED, INC
Danger

A111170-1G A111170-250MG A111170-25G A111170-5G
76687-590EA 23 USD InStock
76687-590 76687-592 76687-594 76687-596
5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid 95%
5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
MDL Number: MFCD03989642
CAS Number: 89001-57-0
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty