Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diammonium oxalate 98%

Supplier: AMBEED, INC
Warning

A114144-1000G A114144-100G A114144-25G A114144-500G
76687-704EA 38 USD InStock
76687-704 76687-706 76687-708 76687-710
Diammonium oxalate 98%
Diammonium oxalate
Precaución: for Research Use Only


Formula: C₂H₈N₂O₄
MDL Number: MFCD00013308
CAS Number: 1113-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty