Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-D-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A114287-100G A114287-10G A114287-1G A114287-250MG A114287-25G
76687-712EA 499 USD InStock
76687-712 76687-714 76687-716 76687-718 76687-720
4-Fluoro-D-phenylalanine 98%
4-Fluoro-D-phenylalanine
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₂
MDL Number: MFCD00063063
CAS Number: 18125-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty