Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Phe-OMe.HCl 97%

Supplier: AMBEED, INC
Warning

A114308-100G A114308-10G A114308-25G A114308-5G
76687-722EA 50 USD InStock
76687-722 76687-724 76687-726 76687-728
H-Phe-OMe.HCl 97%
H-Phe-OMe.HCl
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD00012489
CAS Number: 7524-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty