Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicine 95%

Supplier: AMBEED, INC
Warning

A114701-1000G A114701-100G A114701-10G A114701-25G A114701-500G
76687-742EA 74 USD InStock
76687-742 76687-744 76687-746 76687-748 76687-750
Bicine 95%
Bicine
Precaución: for Research Use Only


Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
MDL Number: MFCD00004295
CAS Number: 150-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty