Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(4-aminophenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A115827-10G A115827-1G A115827-250MG A115827-25G A115827-5G
76687-808EA 17 USD InStock
76687-808 76687-810 76687-812 76687-814 76687-816
Boc-(4-aminophenyl)acetic acid 97%
Boc-(4-aminophenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₄
MDL Number: MFCD00099367
CAS Number: 81196-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty