Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methoxy-5-(methylsulfonyl)benzoate 97%

Supplier: AMBEED, INC
Warning

A117627-100G A117627-10G A117627-1G A117627-25G A117627-5G
76687-954EA 89 USD InStock
76687-954 76687-956 76687-958 76687-960 76687-962
Methyl-2-methoxy-5-(methylsulfonyl)benzoate 97%
Methyl-2-methoxy-5-(methylsulfonyl)benzoate


Formula: C₁₀H₁₂O₅S
MDL Number: MFCD01317545
CAS Number: 63484-12-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty