Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1-phenyl-2-pyrazolin-5-one 98%

Supplier: AMBEED, INC
Warning

A119704-1000G A119704-100G A119704-25G A119704-500G A119704-5G
76688-052EA 63 USD InStock
76688-052 76688-054 76688-056 76688-058 76688-060
3-Methyl-1-phenyl-2-pyrazolin-5-one 98%
3-Methyl-1-phenyl-2-pyrazolin-5-one
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
MDL Number: MFCD00003138
CAS Number: 89-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty